3-(Trifluoromethyl)cinnamic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(3-(Trifluoromethyl)phenyl)acrylic acid
PC7663-100G
PC7663-25G
APOSPC7663-100GEA
2070
CZK
APOSPC7663-100G
APOSPC7663-25G
3-(Trifluoromethyl)cinnamic acid 98%
3-(Trifluoromethyl)cinnamic acid
Vzorec:
C₁₀H₇F₃O₂ MW: 216,16 g/mol Bod tání: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004393 CAS číslo: 779-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...