Tisk…

2,3,5-Trifluorobenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC7654-1G PC7654-250MG
APOSPC7654-1GEA 3110 CZK
APOSPC7654-1G APOSPC7654-250MG
2,3,5-Trifluorobenzyl bromide 97%
2,3,5-Trifluorobenzyl bromide
Vzorec: C₇H₄BrF₃
Storage Temperature: Ambient
MDL Number: MFCD00083543
CAS číslo: 226717-83-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR