1-Fluoro-4-(4-iodophenoxy)benzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoro-4'-iododiphenyl ether
PC7649-1G
PC7649-5G
APOSPC7649-1GEA
3540
CZK
APOSPC7649-1G
APOSPC7649-5G
1-Fluoro-4-(4-iodophenoxy)benzene
1-Fluoro-4-(4-iodophenoxy)benzene
Vzorec:
C₁₂H₈FIO Bod tání: 45…46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368761 CAS číslo: 886762-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...