Tisk…

α'-Chloro-α,α,α-trifluoro-m-xylene 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC7633F-25G PC7633F-100G PC7633F-500G
APOSPC7633F-25GEA 744 CZK
APOSPC7633F-25G APOSPC7633F-100G APOSPC7633F-500G
α'-Chloro-α,α,α-trifluoro-m-xylene 98%
α'-Chloro-α,α,α-trifluoro-m-xylene

Derivitising agent for detection of ethyleneurea by electron-capture G.C.

Vzorec: C₈H₆ClF₃
Bod varu: 68…70 °C/12mmHg
Hustota: 1,254 g/cm³ (25 °C)
Bod vzplanutí: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000908
CAS číslo: 705-29-3
EINECS: 211-884-4
UN: 2920
ADR: 8+3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR