Tisk…

α'-Bromo-α,α,α-trifluoro-p-xylene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide

PC7633-100G PC7633-25G PC7633-5G
APOSPC7633-100GEA 3110 CZK
APOSPC7633-100G APOSPC7633-25G APOSPC7633-5G
α'-Bromo-α,α,α-trifluoro-p-xylene 98%
α'-Bromo-α,α,α-trifluoro-p-xylene
Vzorec: C₈H₆BrF₃
MW: 239.03
Bod varu: 65…66 °C/5mmHg
Bod tání: 30…31 °C
Hustota: 1.546 g/cm³ (25 °C)
Bod vzplanutí: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS číslo: 402-49-3
EINECS: 206-947-8
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR