Tisk…

α'-Bromo-α,α,α-trifluoro-m-xylene 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC7632-100G PC7632-250G PC7632-25G
APOSPC7632-100GEA 2730 CZK
APOSPC7632-100G APOSPC7632-250G APOSPC7632-25G
α'-Bromo-α,α,α-trifluoro-m-xylene 97%
α'-Bromo-α,α,α-trifluoro-m-xylene
Vzorec: C₈H₆BrF₃
Bod varu: 69 °C/4mmHg
Hustota: 1,565 g/cm³ (25 °C)
Bod vzplanutí: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000395
CAS číslo: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR