2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,6-Difluoro-N-methoxy-N-methylbenzamide
PC7619-1G
PC7619-5G
PC7619-250MG
APOSPC7619-1GEA
5290
CZK
APOSPC7619-1G
APOSPC7619-5G
APOSPC7619-250MG
2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide
2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide
Vzorec:
C₉H₉F₂NO₂ MW: 201,17 g/mol Bod tání: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689758 CAS číslo: 937601-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...