N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Dodavatel: Apollo Scientific
PC7606-10G
PC7606-1G
PC7606-5G
APOSPC7606-10GEA
38350
CZK
APOSPC7606-10G
APOSPC7606-1G
APOSPC7606-5G
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Vzorec:
C₁₀H₁₀ClF₃N₂O₂ Bod tání: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689716 CAS číslo: 937601-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...