α,α,α-Trifluoro-o-toluoyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride
PC7590-100G
PC7590-25G
PC7590-500G
APOSPC7590-100GEA
1620
CZK
APOSPC7590-100G
APOSPC7590-25G
APOSPC7590-500G
α,α,α-Trifluoro-o-toluoyl chloride 98%
α,α,α-Trifluoro-o-toluoyl chloride
Vzorec:
C₈H₄ClF₃O MW: 208,57 g/mol Bod varu: 84…85 °C/16mmHg Bod tání: –22 °C Hustota: 1,416 g/cm³ (25 °C) Bod vzplanutí: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000667 CAS číslo: 312-94-7 EINECS: 206-233-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...