2-Bromo-5-(trifluoromethoxy)phenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC7581-1G
PC7581-5G
APOSPC7581-1GEA
4340
CZK
APOSPC7581-1G
APOSPC7581-5G
2-Bromo-5-(trifluoromethoxy)phenylboronic acid 97%
2-Bromo-5-(trifluoromethoxy)phenylboronic acid
Vzorec:
C₇H₅BBrF₃O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD09475807 CAS číslo: 957034-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...