3-Benzyloxy-4-fluorophenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Benzyloxy)-4-fluorophenylboronic acid
PC7572-1G
PC7572-5G
APOSPC7572-1GEA
4370
CZK
APOSPC7572-1G
APOSPC7572-5G
3-Benzyloxy-4-fluorophenylboronic acid 98%
3-Benzyloxy-4-fluorophenylboronic acid
Vzorec:
C₁₃H₁₂BFO₃ MW: 246,0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09475829 CAS číslo: 957034-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...