α,α,α-Trifluoro-m-toluonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Cyanobenzotrifluoride, m-(Trifluoromethyl)benzonitrile, 3-(Trifluoromethyl)benzonitrile
PC7560-25G
PC7560-5G
PC7560-100G
APOSPC7560-25GEA
622
CZK
APOSPC7560-25G
APOSPC7560-5G
APOSPC7560-100G
α,α,α-Trifluoro-m-toluonitrile 98%
α,α,α-Trifluoro-m-toluonitrile
Vzorec:
C₈H₄F₃N MW: 171,12 g/mol Bod varu: 189 °C Bod tání: 14,5 °C Hustota: 1,2821 g/cm³ (25 °C) Bod vzplanutí: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001807 CAS číslo: 368-77-4 EINECS: 206-711-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...