3-Fluoro-4-(methylsulphonyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC7551-1G
PC7551-5G
PC7551-25G
APOSPC7551-1GEA
1820
CZK
APOSPC7551-1G
APOSPC7551-5G
APOSPC7551-25G
3-Fluoro-4-(methylsulphonyl)benzoic acid
3-Fluoro-4-(methylsulphonyl)benzoic acid
Vzorec:
C₈H₇FO₄S MW: 218,2000 g/mol Bod tání: 212…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743730 CAS číslo: 185945-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...