1,1,1,2,2-Pentafluoro-4-iodobutane 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Iodo-3,3,4,4,4-pentafluorobutane
PC7548-100G
PC7548-25G
APOSPC7548-100GEA
7710
CZK
APOSPC7548-100G
APOSPC7548-25G
1,1,1,2,2-Pentafluoro-4-iodobutane 97%
1,1,1,2,2-Pentafluoro-4-iodobutane
Vzorec:
C₄H₄F₅I Bod varu: 100…101 °C Hustota: 1,936 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00155910 CAS číslo: 40723-80-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...