Tisk…

3-Chloro-2-fluorothioanisole 98%

Dodavatel: Apollo Scientific
Varování

PC7535-1G PC7535-5G
APOSPC7535-1GEA 4370 CZK
APOSPC7535-1G APOSPC7535-5G
3-Chloro-2-fluorothioanisole 98%
3-Chloro-2-fluorothioanisole
Vzorec: C₇H₆ClFS
Storage Temperature: Ambient
MDL Number: MFCD09475846
CAS číslo: 214057-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR