α,α,α-Trifluoro-m-toluic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Trifluoromethyl)benzoic acid
PC7530-100G
PC7530-500G
PC7530-1KG
APOSPC7530-100GEA
622
CZK
APOSPC7530-100G
APOSPC7530-500G
APOSPC7530-1KG
α,α,α-Trifluoro-m-toluic acid 98%
α,α,α-Trifluoro-m-toluic acid
Vzorec:
C₈H₅F₃O₂ MW: 190,12 g/mol Bod varu: 237…240 °C Bod tání: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002519 CAS číslo: 454-92-2 EINECS: 207-230-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...