α,α,α-Trifluoro-o-toluic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Trifluoromethyl)benzoic acid
PC7520-100G
PC7520-1KG
PC7520-500G
APOSPC7520-100GEA
992
CZK
APOSPC7520-100G
APOSPC7520-1KG
APOSPC7520-500G
α,α,α-Trifluoro-o-toluic acid 98%
α,α,α-Trifluoro-o-toluic acid
Vzorec:
C₈H₅F₃O₂ MW: 190,12 g/mol Bod varu: 247…254 °C Bod tání: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002476 CAS číslo: 433-97-6 EINECS: 207-093-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...