Tisk…

3-Fluoro-5-methylanisole 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Fluoro-3-methoxy-5-methylbenzene

PC7519-5G PC7519-1G
APOSPC7519-5GEA 2980 CZK
APOSPC7519-5G APOSPC7519-1G
3-Fluoro-5-methylanisole 99%
3-Fluoro-5-methylanisole
Vzorec: C₈H₉FO
Storage Temperature: Ambient
MDL Number: MFCD13185608
CAS číslo: 160911-11-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR