Tisk…

α,α,α-Trifluoro-4-tolualdehyde 98%

Dodavatel: Apollo Scientific

PC7502-100G PC7502-25G
APOSPC7502-100GEA 1820 CZK
APOSPC7502-100G APOSPC7502-25G
α,α,α-Trifluoro-4-tolualdehyde 98%
Vzorec: C₈H₅F₃O
MDL Number: MFCD00006952
CAS číslo: 455-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR