Tisk…

Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate

PC7478-1G PC7478-5G
APOSPC7478-1GEA 4370 CZK
APOSPC7478-1G APOSPC7478-5G
Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Vzorec: C₇H₅BrF₃NO₂S
MW: 304,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08166749
CAS číslo: 72850-79-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR