2-Fluoro-5-methylsulphonyl-3-nitrotoluene
Dodavatel: Apollo Scientific
PC7476-1G
PC7476-5G
APOSPC7476-1GEA
4010
CZK
APOSPC7476-1G
APOSPC7476-5G
2-Fluoro-5-methylsulphonyl-3-nitrotoluene
2-Fluoro-5-methylsulphonyl-3-nitrotoluene
Vzorec:
C₈H₈FNO₄S MW: 233,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09264540 CAS číslo: 1000339-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...