3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,5-Bis(trifluoromethyl)-1,2-diaminobenzene
PC7463-25G
PC7463-5G
APOSPC7463-25GEA
16800
CZK
APOSPC7463-25G
APOSPC7463-5G
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine
Vzorec:
C₈H₆F₆N₂ MW: 244,14 g/mol Bod tání: 42…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631430 CAS číslo: 367-65-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...