1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
Dodavatel: Apollo Scientific
PC7457-1G
PC7457-5G
APOSPC7457-1GEA
3150
CZK
APOSPC7457-1G
APOSPC7457-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
Vzorec:
C₁₂H₁₁F₃N₂O₄ Bod tání: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD01420274 CAS číslo: 1008859-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...