Tisk…

4-Azepan-1-yl-3-nitrobenzotrifluoride

Dodavatel: Apollo Scientific

PC7446-10G PC7446-1G PC7446-5G
APOSPC7446-10GEA 8630 CZK
APOSPC7446-10G APOSPC7446-1G APOSPC7446-5G
4-Azepan-1-yl-3-nitrobenzotrifluoride
4-Azepan-1-yl-3-nitrobenzotrifluoride
Vzorec: C₁₃H₁₅F₃N₂O₂
Bod tání: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00592425
CAS číslo: 62054-71-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR