Tisk…

α-Chloro-4-(trifluoromethoxy)toluene 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Chloromethyl)-4-(trifluoromethoxy)benzene, 4-(Trifluoromethoxy)benzyl chloride, p-(Trifluoromethoxy)benzyl chloride, 1-(Chlormethyl)-4-(trifluormethoxy)benzene

PC7438UF-25G PC7438UF-5G PC7438UF-100G
APOSPC7438UF-25GEA 744 CZK
APOSPC7438UF-25G APOSPC7438UF-5G APOSPC7438UF-100G
α-Chloro-4-(trifluoromethoxy)toluene 97%
α-Chloro-4-(trifluoromethoxy)toluene
Vzorec: C₈H₆ClF₃O
Bod varu: 72…73 °C/15mmHg
Hustota: 1,34 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052326
CAS číslo: 65796-00-1
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR