2-(Trifluoromethoxy)benzyl alcohol 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC7438SB-1G
PC7438SB-25G
PC7438SB-5G
APOSPC7438SB-1GEA
622
CZK
APOSPC7438SB-1G
APOSPC7438SB-25G
APOSPC7438SB-5G
2-(Trifluoromethoxy)benzyl alcohol 97%
2-(Trifluoromethoxy)benzyl alcohol
Vzorec:
C₈H₇F₃O₂ MW: 192.14 Bod varu: 96…98 °C/12mmHg Hustota: 1.33 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00153285 CAS číslo: 175278-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...