Tisk…

3-(Trifluoromethoxy)benzylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC7438O-1G PC7438O-25G PC7438O-5G
APOSPC7438O-1GEA 622 CZK
APOSPC7438O-1G APOSPC7438O-25G APOSPC7438O-5G
3-(Trifluoromethoxy)benzylamine 97%
3-(Trifluoromethoxy)benzylamine
Vzorec: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061267
CAS číslo: 93071-75-1
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR