Tisk…

2'-(Trifluoromethoxy)acetanilide 97%

Dodavatel: Apollo Scientific

PC7433B-25G PC7433B-5G
APOSPC7433B-25GEA 4370 CZK
APOSPC7433B-25G APOSPC7433B-5G
2'-(Trifluoromethoxy)acetanilide 97%
2'-(Trifluoromethoxy)acetanilide
Vzorec: C₉H₈F₃NO₂
Bod tání: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00204176
CAS číslo: 3832-55-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR