Tisk…

1-(Trifluoromethanesulphonyl)imidazole 97%

Dodavatel: Apollo Scientific

Synonyms: 1-((Trifluoromethyl)sulphonyl)-1H-imidazole

PC7433-1G PC7433-25G PC7433-5G
APOSPC7433-1GEA 2900 CZK
APOSPC7433-1G APOSPC7433-25G APOSPC7433-5G
1-(Trifluoromethanesulphonyl)imidazole 97%
1-(Trifluoromethanesulphonyl)imidazole
Vzorec: C₄H₃F₃N₂O₂S
MW: 200,14 g/mol
Bod tání: 19…21 °C
Hustota: 1,525 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037891
CAS číslo: 29540-81-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR