Tisk…

Trifluoromethanesulphonic anhydride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Triflic anhydride

PC7420-250G PC7420-1KG PC7420-500G
APOSPC7420-250GEA 3610 CZK
APOSPC7420-250G APOSPC7420-1KG APOSPC7420-500G
Trifluoromethanesulphonic anhydride 99%
Trifluoromethanesulphonic anhydride
Vzorec: C₂F₆O₅S₂
MW: 282,14 g/mol
Bod varu: 81…83 °C
Bod tání: –80 °C
Hustota: 1,677 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000408
CAS číslo: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR