Tisk…

3-Bromo-5-fluorotoluene 98%

Dodavatel: Apollo Scientific
Varování

PC7412-100G PC7412-25G PC7412-5G
APOSPC7412-100GEA 3410 CZK
APOSPC7412-100G APOSPC7412-25G APOSPC7412-5G
3-Bromo-5-fluorotoluene 98%
3-Bromo-5-fluorotoluene
Vzorec: C₇H₆BrF
MW: 189,03 g/mol
Bod varu: 63…66 °C/2mmHg
Storage Temperature: Ambient
MDL Number: MFCD01861195
CAS číslo: 202865-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR