Trifluoromethanesulphonamide 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC7406-25G
PC7406-5G
PC7406-100G
APOSPC7406-25GEA
4240
CZK
APOSPC7406-25G
APOSPC7406-5G
APOSPC7406-100G
Trifluoromethanesulphonamide 98%
Trifluoromethanesulphonamide
Vzorec:
CH₂F₃NO₂S Bod tání: 119…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068714 CAS číslo: 421-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...