2,2,2-Trifluoroethyl methacrylate 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Methacrylic acid 2,2,2-trifluoroethyl ester
PC7374-100G
PC7374-25G
PC7374-500G
APOSPC7374-100GEA
1210
CZK
APOSPC7374-100G
APOSPC7374-25G
APOSPC7374-500G
2,2,2-Trifluoroethyl methacrylate 99%
2,2,2-Trifluoroethyl methacrylate
Vzorec:
C₆H₇F₃O₂ MW: 168,12 g/mol Bod varu: 29…30 °C/40mmHg Hustota: 1,181 g/cm³ (25 °C) Bod vzplanutí: 16 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013576 CAS číslo: 352-87-4 EINECS: 206-525-3 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...