2',3',5'-Trifluoroacetophenone 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(2,3,5-Trifluorophenyl)ethanone, 1-(2,3,5-Trifluorophenyl)ethan-1-one
PC7346-10G
PC7346-5G
PC7346-1G
PC7346-25G
APOSPC7346-10GEA
8200
CZK
APOSPC7346-10G
APOSPC7346-5G
APOSPC7346-1G
APOSPC7346-25G
2',3',5'-Trifluoroacetophenone 98%
2',3',5'-Trifluoroacetophenone
Vzorec:
C₈H₅F₃O MW: 174,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00083548 CAS číslo: 243459-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...