1-([4-Nitro-3-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid
Dodavatel: Apollo Scientific
PC7333-1G
PC7333-500MG
APOSPC7333-1GEA
11770
CZK
APOSPC7333-1G
APOSPC7333-500MG
1-([4-Nitro-3-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid
1-([4-Nitro-3-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid
Vzorec:
C₁₃H₁₃F₃N₂O₄ MW: 318,25 g/mol Bod tání: 153…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD08704869 CAS číslo: 942474-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...