2,3,6-Trifluorobenzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
α-Bromo-2,3,6-trifluorotoluene, 2-(Bromomethyl)-1,3,4-trifluorobenzene
PC7285P-25G
PC7285P-5G
APOSPC7285P-25GEA
5390
CZK
APOSPC7285P-25G
APOSPC7285P-5G
2,3,6-Trifluorobenzyl bromide 97%
2,3,6-Trifluorobenzyl bromide
Vzorec:
C₇H₄BrF₃ MW: 225,01 g/mol Bod varu: 114 °C Hustota: 1,718 g/cm³ (25 °C) Bod vzplanutí: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00061208 CAS číslo: 151412-02-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...