2,3,4-Trifluorobenzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene
PC7285N-1G
PC7285N-5G
PC7285N-25G
APOSPC7285N-1GEA
622
CZK
APOSPC7285N-1G
APOSPC7285N-5G
APOSPC7285N-25G
2,3,4-Trifluorobenzyl bromide 97%
2,3,4-Trifluorobenzyl bromide
Vzorec:
C₇H₄BrF₃ MW: 225,01 g/mol Hustota: 1,71 g/cm³ (25 °C) Bod vzplanutí: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061233 CAS číslo: 157911-55-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...