Tisk…

2,4,6-Trifluorobenzoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC7285H-5G PC7285H-25G PC7285H-100G
APOSPC7285H-5GEA 622 CZK
APOSPC7285H-5G APOSPC7285H-25G APOSPC7285H-100G
2,4,6-Trifluorobenzoyl chloride 98%
2,4,6-Trifluorobenzoyl chloride
Vzorec: C₇H₂ClF₃O
Bod varu: 167…168 °C
Hustota: 1,499 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070810
CAS číslo: 79538-29-7
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR