Tisk…

2-(Difluoromethoxy)iodobenzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(Difluoromethoxy)-2-iodobenzene

PC7263-25G PC7263-5G
APOSPC7263-25GEA 4370 CZK
APOSPC7263-25G APOSPC7263-5G
2-(Difluoromethoxy)iodobenzene
2-(Difluoromethoxy)iodobenzene
Vzorec: C₇H₅F₂IO
MW: 270.0153 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09749662
CAS číslo: 920981-12-0
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR