Perfluoro-3,6-dioxadecanoic acid
Dodavatel: Apollo Scientific
PC7256-100G
PC7256-25G
PC7256-1G
PC7256-5G
APOSPC7256-100GEA
0
CZK
APOSPC7256-100G
APOSPC7256-25G
APOSPC7256-1G
APOSPC7256-5G
Perfluoro-3,6-dioxadecanoic acid
Perfluoro-3,6-dioxadecanoic acid
Vzorec:
C₈HF₁₅O₄ Bod varu: 55 °C/6mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03094421 CAS číslo: 137780-69-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...