Tisk…

4-Bromo-1-fluoro-2-naphthaleneboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Bromo-1-fluoronaphthalen-2-yl)boronic acid

PC7247-1G PC7247-5G
APOSPC7247-1GEA 4200 CZK
APOSPC7247-1G APOSPC7247-5G
4-Bromo-1-fluoro-2-naphthaleneboronic acid 98%
4-Bromo-1-fluoro-2-naphthaleneboronic acid
Vzorec: C₁₀H₇BBrFO₂
MW: 268,8748 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027236
CAS číslo: 913836-09-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR