Tisk…

2-Chloro-3,6-difluoroanisole 98%

Dodavatel: Apollo Scientific

PC7205-5G PC7205-1G PC7205-25G
APOSPC7205-5GEA 2900 CZK
APOSPC7205-5G APOSPC7205-1G APOSPC7205-25G
2-Chloro-3,6-difluoroanisole 98%
2-Chloro-3,6-difluoroanisole
Vzorec: C₇H₅ClF₂O
MW: 178.57
Storage Temperature: Ambient
MDL Number: MFCD01631451
CAS číslo: 261762-37-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR