Tisk…

2-Fluoro-5-hydroxybenzenecarbonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Fluoro-5-hydroxybenzonitrile

PC7202-1G PC7202-5G
APOSPC7202-1GEA 870 CZK
APOSPC7202-1G APOSPC7202-5G
2-Fluoro-5-hydroxybenzenecarbonitrile 98%
2-Fluoro-5-hydroxybenzenecarbonitrile
Vzorec: C₇H₄FNO
MW: 137,1100 g/mol
Bod tání: 98…108 °C
Storage Temperature: Ambient
MDL Number: MFCD11846514
CAS číslo: 104798-53-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR