Tisk…

α,α,α-Trifluoroacetophenone 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

PC7200-100G PC7200-25G PC7200-5G
APOSPC7200-100GEA 3480 CZK
APOSPC7200-100G APOSPC7200-25G APOSPC7200-5G
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Vzorec: C₈H₅F₃O
MW: 174,12 g/mol
Bod varu: 165…166 °C
Hustota: 1,24 g/cm³ (25 °C)
Bod vzplanutí: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS číslo: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR