Tisk…

4-Amino-2-methyl-8-(trifluoromethyl)quinoline

Dodavatel: Apollo Scientific

PC7198-1G PC7198-500MG
APOSPC7198-1GEA 9530 CZK
APOSPC7198-1G APOSPC7198-500MG
4-Amino-2-methyl-8-(trifluoromethyl)quinoline
4-Amino-2-methyl-8-(trifluoromethyl)quinoline
Vzorec: C₁₁H₉F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD08669720
CAS číslo: 147147-73-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR