Tisk…

3-Methyl-5-(trifluoromethoxy)benzyl alcohol 97%

Dodavatel: Apollo Scientific

PC7192-10G PC7192-5G
APOSPC7192-10GEA 10350 CZK
APOSPC7192-10G APOSPC7192-5G
3-Methyl-5-(trifluoromethoxy)benzyl alcohol 97%
3-Methyl-5-(trifluoromethoxy)benzyl alcohol
Vzorec: C₉H₉F₃O₂
MW: 206,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025373
CAS číslo: 916420-54-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR