1,1,1-Trifluoroacetone cyanohydrin
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1,1,1-Trifluoroacetone Cyanohydrin, 2-Cyano-1,1,1-trifluoro-2-propanol, 2-Hydroxy-2-(trifluoromethyl)propionitrile
PC7185-1G
PC7185-5G
APOSPC7185-1GEA
6850
CZK
APOSPC7185-1G
APOSPC7185-5G
1,1,1-Trifluoroacetone cyanohydrin
1,1,1-Trifluoroacetone cyanohydrin
Vzorec:
C₄H₄F₃NO MW: 139,08 g/mol Bod varu: 68…69 °C/40mmHg Hustota: 1,276 g/cm³ (25 °C) Bod vzplanutí: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040885 CAS číslo: 335-08-0 UN: 2929 ADR: 6.1+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...