Tisk…

2-Bromo-6-fluorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Bromo-6-fluorophenyl)boronic acid, 2-Bromo-6-fluorobenzeneboronic acid

PC7182-100G PC7182-25G PC7182-5G
APOSPC7182-100GEA 15340 CZK
APOSPC7182-100G APOSPC7182-25G APOSPC7182-5G
2-Bromo-6-fluorophenylboronic acid 98%
2-Bromo-6-fluorophenylboronic acid
Vzorec: C₆H₅BBrFO₂
MW: 218,82 g/mol
Bod tání: 138…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664309
CAS číslo: 913835-80-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR