3-(Trifluoromethyl)-5-pyridineboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid
PC7177-1G
APOSPC7177-1GEA
3440
CZK
APOSPC7177-1G
3-(Trifluoromethyl)-5-pyridineboronic acid 95%
3-(Trifluoromethyl)-5-pyridineboronic acid
Vzorec:
C₆H₅BF₃NO₂ MW: 190,9156 g/mol Bod tání: 260…262,6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09952041 CAS číslo: 947533-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...