Tisk…

3-Cyano-5-fluorophenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Cyano-5-fluorophenyl)boronic acid

PC7176-1G PC7176-5G
APOSPC7176-1GEA 1370 CZK
APOSPC7176-1G APOSPC7176-5G
3-Cyano-5-fluorophenylboronic acid 97%
3-Cyano-5-fluorophenylboronic acid
Vzorec: C₇H₅BFNO₂
MW: 164,9300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09263608
CAS číslo: 304858-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR